Ac-LEHD-pNA
Proveedor: ENZO LIFE SCIENCES
ALX260081M001 ALX260081M005
ENZOALX260081M001EA 84.7 EUR
ENZOALX260081M001 ENZOALX260081M005
Ac-LEHD-pNA
Proteínas y Péptidos
Caspase-9 substrate
Chromogenic substrate for caspase-9 and cysteine proteases. Similar to Ac-LEHD-AMC (Prod. No. ALX-260-080) and Ac-LEHD-AFC (Prod. No. ALX-260-116) but cleavage is monitored colorimetrically by absorbance at 405 nm.