Human recombinant TNF alpha
Proveedor: ENZO LIFE SCIENCES
ALX520002C010 ALX520002C050
ENZOALX520002C010EA 267 EUR
ENZOALX520002C010 ENZOALX520002C050
Human recombinant TNF alpha
Proteínas y Péptidos
Produced in yeast.
New Product
Order Now
ESPECIFICACIONES
Nombre de proteína / péptido TNF alpha TNF alpha
Environmentally Preferable
Tipo de proteína / péptido Recombinant Recombinant
Especies Human Human
Fuente
Actividad biológica Induces cytolysis of mouse L929 cells in the presence of 1µg/ml Actinomycin D (ED₅₀≤0,05 ng/ml).
Pureza >97% (SDS-PAGE)
Grado Cell culture Grade Cell culture Grade
Formulación Lyophilised from 0,2 μm-filtered solution containing mannitol.
Instrucciones de reconstitución A quick spin is recommended followed by reconstitution in distilled, sterile water to a concentration of 0,1 - 1 mg/ ml. For further dilution, add a carrier protein to avoid loss of biological activity.