Human recombinant alpha-B-Crystallin,HIS tag
Proveedor: ENZO LIFE SCIENCES
ADISPP228050 ADISPP228200
ENZOADISPP228050EA 255 EUR
ENZOADISPP228050 ENZOADISPP228200
Human recombinant alpha-B-Crystallin,HIS tag
Proteínas y Péptidos
Produced in E. coli. Human αB-Crystallin is fused at the N-terminus to a His-tag.