Proteasome inhibitor II aldehyde
Proveedor: ENZO LIFE SCIENCES
ALX260090M005
ENZOALX260090M005EA
308
EUR
ENZOALX260090M005
Proteasome inhibitor II aldehyde
Proteínas y Péptidos
Potent and cell permeable proteasome inhibitor. Inhibits the chymotrypsin-like activity of the 26S proteasome (MCP; multicatalytic proteinase complex). Does not inhibit the peptidylglutamyl-peptide hydrolyzing activity of the MPC.
White to off-white powder.
Soluble in DMSO.