Ac-VEID-AFC
Proveedor: ENZO LIFE SCIENCES
ALX260111M005 ALX260111M010
ENZOALX260111M005EA 304 EUR
ENZOALX260111M005 ENZOALX260111M010
Ac-VEID-AFC
Proteínas y Péptidos
Caspase-6 inhibitor
Fluorogenic substrate for caspase-6 (Km=30 μM; Kcat/Km=168000 M-1sec-1) and related cysteine proteases. Sequence is based on lamin A site of cleavage.