Z-Leu-Leu-CHO
Proveedor: ENZO LIFE SCIENCES
BMLPI1160005 BMLPI1160025
ENZOBMLPI1160005EA 86.9 EUR
ENZOBMLPI1160005 ENZOBMLPI1160025
Z-Leu-Leu-CHO
Proteínas y Péptidos
Calpain inhibitor
Selective inhibitor of calpain, 100-fold selective for calpain over proteasome.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e63027f796d58427e68f33690e3be481622f6c8&vid=f0d786bd6069305f1ab8b58125f30e619416ddb2&prid=web&clid=SAPDAM