PPACK
Proveedor: ENZO LIFE SCIENCES
BMLPI1170005 BMLPI1170025
ENZOBMLPI1170005EA 183 EUR
ENZOBMLPI1170005 ENZOBMLPI1170025
PPACK
Proteínas y Péptidos
Potent irreversible thrombin inhibitor. Inhibits thrombin activation of progelatinase A. Pretreatment with PPACK inhibits thrombin-induced cerebral thromboembolism in vivo.
  • Thrombin inhibitor
White to off-white solid.
Soluble in water.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=907a031f630c2f5f283ed1fcf84f522e3085421c&vid=24d0c6c6ff7700a19eec9c72c38b1a29fe88aead&prid=web&clid=SAPDAM