Disprosio ≥99.9% (REO, rare earth oxide basis), láminas, Grosor: 0.025 mm (0.001 in)
Proveedor: Thermo Fisher Scientific
|
Peligro
|
acondicionado en Argón
12383.FF
12383.FI
12383.FFEA
349
EUR
12383.FF
12383.FI
Disprosio ≥99.9% (REO, rare earth oxide basis), láminas, Grosor: 0.025 mm (0.001 in)
Disprosio
Purity: 99.9%
CAS No.: 7429-91-6
Physical Form: ≈0.13g/25x25mm, Packaged under argon
Appearance: Gray
Odour: Odourless
UN No.: 3178
MDL No.: MFCD00010982
Formula:
Dy MW: 162.5 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00010982 Núm. CAS: 7429-91-6 EINECS: 231-073-9 UN: 3178 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...