Coloring reagent for nitrites dosage SKALAR
Proveedor: PROLABO CMP
|
Atención
|
MC310971.1000
MC310971.5000
PRLSMC310971.1000EA
0
EUR
PRLSMC310971.1000
PRLSMC310971.5000
Coloring reagent for nitrites dosage SKALAR
Coloring reagent for nitrites dosage
Densidad:
0,98 g/cm³ (20 °C) |
EINECS:
264-151-6 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...