D(+)-Triptofano ≥99%, en polvo
Proveedor: MP Biomedicals
Sinónimos:
H-D-Trp-OH
0210314425
0210314480
0210314401
ICNA0210314425EA
172
EUR
ICNA0210314425
ICNA0210314480
ICNA0210314401
D(+)-Triptofano ≥99%, en polvo
D(+)-Triptofano
D-Tryptophan is an amino acid and a unnatural isomer of tryptophan.
Soluble in water or hot alcohol. Also soluble in 0,5 M Hydrochloric Acid (50 mg plus 1 ml 0,5 M HCl) giving clear to slightly hazy solution.
Formula:
C₁₁H₁₂N₂O₂ MW: 204,23 g/mol Pto. de fusión: 280…283 °C |
Número MDL:
MFCD00005647 Núm. CAS: 153-94-6 EINECS: 205-819-9 Índice Merck: 14,09797 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...