Alcohol isoamílico ≥98.5% (por GC), Puriss. p.a. ACS, Riedel-de Haën™

Proveedor: Honeywell Chemicals
Atención

59090-1L 59090-200L
59090-1LEA 88.9 EUR
59090-1L HONC59090-200L
Alcohol isoamílico ≥98.5% (por GC), Puriss. p.a. ACS, Riedel-de Haën™
Alcohol isoamílico

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 1718835
RTECS Number: EL5425000
UNSPSC Code: 12190000

Formula: (CH₃)₂CHCH₂CH₂OH
Pto. Ebullición: 131-132 °C
Pto. de fusión: -117 °C
Densidad: 0,81 g/cm³ (20 °C)
Pto. de inflamación: 43 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00002934
Núm. CAS: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3,III
Índice Merck: 13,05214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=170c60e26c7b4edab6e0686f91d04e676da16e6d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Identity (IR) complying
Assay (GC) Min. 98.5 %
Refractive index (n 20/D) 1.406 - 1.408
Non-volatile matter Max. 0.003 %
Water (Karl Fischer) Max. 0.3 %
Free acid (as CH₃COOH) Max. 0.01 %
Acids, esters (as amyl acetate) Max. 0.06 %
Titrable Acid Max. 0.002 meq/g
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 2 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
2-methylbutanol (GC) Max. 0.1 %
Carbonyl Compounds (as HCHO) Max. 0.1 %

ESPECIFICACIONES

Environmentally Preferable Safer Human and Environmental Health
Certificación ambiental Chem21 Solvent Selection Guide recommended by default

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR