Ácido (2,3-diclorofenil)acético ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2,3-Dichlorophenylacetic acid

D2931-25G D2931-5G
TCIAD2931-25GEA 223 EUR
TCIAD2931-25G TCIAD2931-5G
Ácido (2,3-diclorofenil)acético ≥98.0% (by GC, titration analysis)
Ácido (2,3-diclorofenil)acético
Formula: (C₆H₃Cl₂)CH₂COOH
MW: 205,04 g/mol
Número MDL: MFCD01861393
Núm. CAS: 10236-60-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dabf41e5c219ea67ad438c86f60350d52afff36&vid=5dabf41e5c219ea67ad438c86f60350d52afff36&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 132.0 to 136.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR