3-Buten-1-ol 98+%
Proveedor: Thermo Fisher Scientific
Sinónimos:
1-Buten-4-ol
295750250
295751000
295755000
ACRO295750250EA
142
EUR
ACRO295750250
ACRO295751000
ACRO295755000
3-Buten-1-ol 98+%
3-Buten-1-ol
Formula:
C₄H₈O MW: 72.11 g/mol |
Número MDL:
MFCD00002959 Núm. CAS: 627-27-0 |
Resultados test especificaciones
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4200 to 1.4220 (20°C, 589 nm) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...