Bromuro de calcio, anhidro ≥99.5%
Proveedor: Thermo Fisher Scientific
|
Atención
|
87893.09
87893.18
87893.30
87893.09EA
34.2
EUR
87893.09
87893.18
87893.30
Bromuro de calcio, anhidro ≥99.5%
Bromuro de calcio
Purity: 99.5%
CAS No.: 7789-41-5
Molecular Formula: CaBr2
Formula Weight: 199.90
Physical Form: -50 Mesh Powder
Density: 3.353
Appearance: White to off-white
Odour: Odourless
Melting Point: 730°
Boiling Point: 806-812°
Solubility: Freely soluble in water, methanol, ethanol. Soluble in acetone. Insoluble in dioxane, chloroform, ether
Merck Reference: 14,1654
MDL No.: MFCD00010902
Photography, dehydrating agent, manufacturing of mineral water, fire retardant, wood preservative, freezing mixture, road treatment
Formula:
Br₂Ca Pto. Ebullición: 806…812 °C Pto. de fusión: 730 °C Densidad: 3,353 Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00010902 Núm. CAS: 7789-41-5 EINECS: 232-164-6 Índice Merck: 14,1654 |