Diclorodietilgermano
Proveedor: Thermo Fisher Scientific
|
Peligro
|
CAS No.: 13314-52-8
Molecular Formula: (C2H5)2GeCl2
Formula Weight: 201.64
Flash Point: 63°(145°F)
Physical Form: Liquid
Density: 1.372
Refractive Index: 1.4710
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Acrid
Melting Point: -37°
Boiling Point: 175°
Solubility: Decomposes in water and NH3. Soluble in organic solvents
UN No.: 3265
MDL No.: MFCD00013589
As an intermediate
Formula:
C₄H₁₀Cl₂Ge Pto. Ebullición: 175 °C Pto. de fusión: –37 °C Densidad: 1,372 Pto. de inflamación: 63 °C (145 °F) Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00013589 Núm. CAS: 13314-52-8 EINECS: 236-344-5 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...