Tetrafluoroborato de amonio ≥99.5% (metals basis)

Proveedor: Thermo Scientific
Peligro

Sinónimos: Ammonium fluoroborate, ammonium fluoborate ammonium borofluoride ammonium fluoroborate [qr] ammonium tetrafluoroborate

35831.30 35831.A1
35831.30EA 112 EUR
35831.30 35831.A1
Tetrafluoroborato de amonio ≥99.5% (metals basis)
Ammonium tetrafluoroborate

Purity: 99.5% CAS No.: 13826-83-0 Molecular Formula: NH4BF4 Formula Weight: 104.84 Physical Form: -20 Mesh Density: 1.871 Appearance: White Odour: Ammonia-like Melting Point: 220° subl. UN No.: 3260 MDL No.: MFCD06245785

Formula: H₄BF₄N
MW: 104,84 g/mol
Pto. de fusión: 220 °C (subl.)
Densidad: 1,871
Temperatura de almacenaje: Ambiente
Número MDL: MFCD06245785
Núm. CAS: 13826-83-0
EINECS: 237-531-4
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR