Tetrafluoroborato de amonio ≥99.999% (metals basis)
Proveedor: Thermo Fisher Scientific
|
Peligro
|
Sinónimos:
Ammonium fluoroborate, ammonium fluoborate ammonium borofluoride ammonium fluoroborate [qr] ammonium tetrafluoroborate
44055.06
44055.14
44055.22
44055.06EA
43
EUR
44055.06
44055.14
44055.22
Tetrafluoroborato de amonio ≥99.999% (metals basis)
Tetrafluoroborato de amonio
Purity: 99.999%
CAS No.: 13826-83-0
Molecular Formula: NH4BF4
Formula Weight: 104.84
Physical Form: Crystalline
Density: 1.871
Appearance: White
Odour: Ammonia-like
Melting Point: 220° subl.
UN No.: 3260
MDL No.: MFCD06245785
Formula:
H₄BF₄N Pto. de fusión: 220 °C (subl.) Densidad: 1,871 Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD06245785 Núm. CAS: 13826-83-0 EINECS: 237-531-4 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...