HPLC columns, ZORBAX® HILIC Plus
Proveedor: Agilent
959793-901 959961-901 959743-901 959943-901
HEWL959793-901EA 755 EUR
HEWL959793-901 HEWL959961-901 HEWL959743-901 HEWL959943-901
HPLC columns, ZORBAX® HILIC Plus
Columnas de cromatografía
HILIC Plus columns are designed for hydrophilic interaction chromatography (HILIC) applications - typically used for the retention and resolution of small polar compounds.
  • High sensitivity for LC/MS applications
  • Non-bonded silica columns
  • Silica provides excellent peak shape, critical for many polar, basic analytes
  • Mobile phase: 90% acetontrile and 10% water
HILIC Plus is very similar to InfinityLab Poroshell 120 HILIC.