Safety shoes, lace-up, Basic, Lenny S2, Jori
Proveedor: ELTEN
12320/36 12320/37 12320/38 12320/39 12320/40 12320/41 12320/42 12320/43 12320/44 12320/45 12320/46 12320/47 12320/48
ELTN12320/36EA 38.9 EUR
ELTN12320/36 ELTN12320/37 ELTN12320/38 ELTN12320/39 ELTN12320/40 ELTN12320/41 ELTN12320/42 ELTN12320/43 ELTN12320/44 ELTN12320/45 ELTN12320/46 ELTN12320/47 ELTN12320/48
Safety shoes, lace-up, Basic, Lenny S2, Jori
Zapatos y botas
Durable safety shoes made from wear resistant cowhide leather, skin friendly textile lining, a full padded bellows tongue and collar padding plus a dual-density PU/PU New Basic sole (oil, fuel and abrasion resistant, also heat resistant up to approx. 130 °C).
  • Climate-regulating, breathable textile lining
  • Full length inlay sole
  • Anti-static soft fleece insole
  • PU scuff cap protects the upper material
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: Black
Certificaciones: EN ISO 20345; S2, SRC