Perclorato de plata, anhidro
Proveedor: Thermo Fisher Scientific
|
Peligro
|
42209.06
42209.14
42209.22
42209.06EA
58.5
EUR
42209.06
42209.14
42209.22
Perclorato de plata, anhidro
Perclorato de plata
CAS No.: 7783-93-9
Molecular Formula: AgClO4
Formula Weight: 207.32
Physical Form: Crystalline
Density: 2.806
Sensitivity: Light Sensitive and Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 485° dec.
Solubility: Soluble in water and many organic solvents (benzene, toluene, pyridine)
Merck Reference: 14,8523
UN No.: 1481
MDL No.: MFCD00003400
Formula:
AgClO₄ MW: 207.32 g/mol Pto. de fusión: 485 °C (desc.) Densidad: 2.806 Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00003400 Núm. CAS: 7783-93-9 EINECS: 232-035-4 UN: 1481 ADR: 5.1 II Índice Merck: 14,8523 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...