Sulfuro de cobre ≥99.8% (metals basis)
Proveedor: Thermo Fisher Scientific
Sinónimos:
Copper sulphide, Cupric sulphide, Copper(II)sulphide
42925.18
42925.30
42925.A1
42925.18EA
124
EUR
42925.18
42925.30
42925.A1
Sulfuro de cobre ≥99.8% (metals basis)
Sulfuro de cobre
Purity: 99.8%
CAS No.: 1317-40-4
Molecular Formula: CuS
Formula Weight: 95.60
Physical Form: -200 Mesh Powder
Density: 3.9-4.6
Refractive Index: 1.45
Sensitivity: Air and Moisture Sensitive
Melting Point: 220° dec.
Merck Reference: 14,2655
MDL No.: MFCD00016066
Formula:
CuS Pto. de fusión: 220 °C (desc.) Densidad: 3,9…4.6 Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00016066 Núm. CAS: 1317-40-4 EINECS: 215-271-2 Índice Merck: 14,2655 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...