Pentacloruro de tántalo ≥99.8% (metals basis)
Proveedor: Thermo Scientific
|
Peligro
|
Ampuled under argon
14164.06
14164.14
14164.22
14164.06EA
65.2
EUR
14164.06
14164.14
14164.22
Pentacloruro de tántalo ≥99.8% (metals basis)
Pentacloruro de tántalo
Purity: 99.8%
CAS No.: 7721-01-9
Molecular Formula: TaCl5
Formula Weight: 358.21
Physical Form: Powder, Ampouled under argon
Density: 3.68
Appearance: White to pale yellow
Odour: Acidic
Melting Point: 216°
Boiling Point: 242°
Solubility: Decomposes in water. Soluble in absolute alcohol and KOH
Merck Reference: 14,9054
UN No.: 3260
MDL No.: MFCD00011253
In chlorination of organic substances, as an intermediate, and in the production of pure metal
Formula:
Cl₅Ta MW: 358,21 g/mol Pto. Ebullición: 242 °C Pto. de fusión: 216 °C Densidad: 3,68 Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00011253 Núm. CAS: 7721-01-9 EINECS: 231-755-6 UN: 3260 ADR: 8,III Índice Merck: 14,9054 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...