6-Bencilaminopurina ≥99%
Proveedor: Thermo Scientific
|
Atención
|
Sinónimos:
Bencil(purin-6-il)amina
A14678.03
A14678.06
A14678.14
A14678.03EA
32.5
EUR
A14678.03
A14678.06
A14678.14
6-Bencilaminopurina ≥99%
6-Bencilaminopurina
Purity: 99%
CAS No.: 1214-39-7
Molecular Formula: C12H11N5
Formula Weight: 225.26
Melting Point: 229-233°
Beilstein Reference: 19406
Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables
Formula:
C₁₂H₁₁N₅ MW: 225,25 g/mol Pto. de fusión: 229…233 °C Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00005572 Núm. CAS: 1214-39-7 EINECS: 214-927-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...