Capillary columns, GS-GasPro
Proveedor: Agilent
113-4302 113-4312 113-4332 113-4362
HEWL113-4302EA 455 EUR
HEWL113-4302 HEWL113-4312 HEWL113-4332 HEWL113-4362
Capillary columns, GS-GasPro
Columnas de cromatografía
These columns are ideal for GC/MS.
  • Excellent choice for light hydrocarbons and sulphur gases, and can separate CO and CO₂ on a single column
  • Extended temperature range of – 80 to 260/300 °C
  • High levels of retention stability that is not affected by water
Unique bonded PLOT column technology.