Winter safety ankle boots, lace-up/Zip, Basic, Jerry, Jori
Proveedor: ELTEN
16011/36 16011/37 16011/38 16011/39 16011/40 16011/41 16011/42 16011/43 16011/44 16011/45 16011/46 16011/47 16011/48
ELTN16011/36EA 55.9 EUR
ELTN16011/36 ELTN16011/37 ELTN16011/38 ELTN16011/39 ELTN16011/40 ELTN16011/41 ELTN16011/42 ELTN16011/43 ELTN16011/44 ELTN16011/45 ELTN16011/46 ELTN16011/47 ELTN16011/48
Winter safety ankle boots, lace-up/Zip, Basic, Jerry, Jori
Zapatos y botas
Warm winter safety boot feature a zip/lace-up combination that are made from cowhide leather, warm winter lining, a full padded bellows tongue, collar padding plus a dual-density PU/PU jo_Basic sole (oil, fuel and abrasion resistant, also heat resistant up to approx. 130 °C).
  • Warm, moisture-absorbent lining
  • Full length inlay sole
  • Anti-static soft fleece insole
  • PU scuff cap protects the upper material
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificaciones: EN ISO 20345; S3, SRC