Fmoc-Gly-(Dmb)Gly-OH

Proveedor: Novabiochem (Part of Merck)

8.52109.0001 8.52109.0005
04-12-1266-1EA 249 EUR
04-12-1266-1 8.52109.0005
Fmoc-Gly-(Dmb)Gly-OH
Proteínas y Péptidos

Fmoc-Aaa-(Dmb)Gly-OH dipeptides offer the same benefits as pseudoproline dipeptides but for peptide sequences containing Gly.


  • White to slight yellow powder
  • Water (K.F.): ≤2,00%
  • Assay: ≥90,0% (acidimetric) and 95,0% (a/a - HPLC)


They are extremely easy to use. Standard coupling methods like PyBOP®/DIPEA or DIPCDI/HOBt can be used for their introduction. Removal of the Dmb group and regeneration of the glycine residue occurs during the course of standard TFA-mediated cleavage reaction. The use of this derivative was found to be essential for the synthesis of peptides related to nucleolin.

Order Now

ESPECIFICACIONES

Nombre de proteína / péptido Fmoc-Gly-(Dmb)Gly-OH
Environmentally Preferable
Sinónimos de proteínas N-α-Fmoc-L-glycyl-N-α-(2|4-dimethoxybenzyl)-glycine
Especies
Fuente
CAS No. 848861-65-4
Pureza >98%
Molecular weight 504.53 g/mol
Molecular formula C₂₈H₂₈N₂O₇
Condiciones de almacenamiento 2...8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR