Professional ventilated refrigerators, GKPv
Proveedor: Liebherr
GKPV6520 GKPV6570 GKPV1470 GKPV6590 GKPV6573
472-1063EA 0 EUR
472-1063 472-1064 472-1060 471-0910 471-1066
Professional ventilated refrigerators, GKPv
Refrigeradores
These commercial refrigerators feature a fan for forced air cooling with temperature adjustable from –2 to +15 °C or +1 to +15 °C.
  • Electronic temperature regulation, with digital display
  • Alarm for temperature and door opening
  • Automatic defrosting with evaporation of condensates
  • Interior with rounded corners to make cleaning easier
  • Door is lockable and reversible, self closing at 45° but stable at 90°
Interior and exterior made from stainless steel AISI 306, with indirect ventilated cooling, insulation 83 mm (allows temperature controlMore Product Information
Order Now
ESPECIFICACIONES
Modelo GKPv 6520 GKPv6570 GKPv1470 GKPv 6590 GKPv 6573
Environmentally Preferable
Referencia 472-1063 472-1064 472-1060 471-0910 471-1066
Temperatura +1...+15 °C +1...+15 °C −2...+16 °C
Pantalla Digital Digital Digital Digital Digital
Descongelación Automatic Automatic Automatic Automatic Automatic
Alarma Audible and visual Visual y auditivo Visual y auditivo Audible and visual Audible and visual
Estantes/cajones 4 plastic-coated shelves 4 estantes 2x4 estantes 4 plastic-coated shelves 4 plastic-coated shelves
Tipo de puerta 1 solid swing door 1 solid swing door 2 solid swing doors 1 solid swing door 1 glazed swing door, with stainless steel frame
Bloqueo de puerta Yes Yes Yes
Especificaciones técnicas Electronic temperature controller Electronic temperature controller Electronic temperature controller Electronic temperature controller, multi-function digital display and RS485 interface Electronic temperature controller
An×F×Al ext. 700×830×2150 mm 700×830×2150 mm 1430×830×2150 mm 700×830×2150 mm 700×830×2150 mm
Peso 118 kg 180 kg 239 kg 180 kg 144 kg