HPLC columns, NUCLEODUR® Phenyl-Hexyl
Proveedor: MACHEREY-NAGEL
760589.46 760566.46 760586.20 760561.46 760566.40 760561.40 760563.20 760563.30 760563.40 760563.46 760565.20 760566.20 760566.30 760568.20 760573.20 760573.46 760575.46 760576.20 760576.30 760576.46 760577.20 760577.30 760577.40 760577.46 760578.20 760578.30 760578.40 760578.46 760579.30 760579.40 760579.46 760583.20 760583.30 760583.46 760586.30 760586.46 760587.30 760587.40 760587.46 760588.20 760588.30 760588.40 760588.46 760589.30 760589.40 761985.20 761985.30 761986.20 761986.30 761987.20 761987.30 760585.46 760587.20 760589.20 760561.20 760576.40 760579.20 760573.30 760583.40 760561.30 760573.40 760586.40
MANA760589.46EA 760 EUR
MANA760589.46 MANA760566.46 MANA760586.20 MANA760561.46 MANA760566.40 MANA760561.40 MANA760563.20 MANA760563.30 MANA760563.40 MANA760563.46 MANA760565.20 MANA760566.20 MANA760566.30 MANA760568.20 MANA760573.20 MANA760573.46 MANA760575.46 MANA760576.20 MANA760576.30 MANA760576.46 MANA760577.20 MANA760577.30 MANA760577.40 MANA760577.46 MANA760578.20 MANA760578.30 MANA760578.40 MANA760578.46 MANA760579.30 MANA760579.40 MANA760579.46 MANA760583.20 MANA760583.30 MANA760583.46 MANA760586.30 MANA760586.46 MANA760587.30 MANA760587.40 MANA760587.46 MANA760588.20 MANA760588.30 MANA760588.40 MANA760588.46 MANA760589.30 MANA760589.40 MANA761985.20 MANA761985.30 MANA761986.20 MANA761986.30 MANA761987.20 MANA761987.30 MANA760585.46 MANA760587.20 MANA760589.20 MANA760561.20 MANA760576.40 MANA760579.20 MANA760573.30 MANA760583.40 MANA760561.30 MANA760573.40 MANA760586.40
HPLC columns, NUCLEODUR® Phenyl-Hexyl
Columnas de cromatografía
NUCLEODUR® Phenyl-Hexyl is a phenyl-hexyl modified silica phase with multi-endcapping for HPLC.
  • High purity spherical silica gel
  • Hydrophobic RP phase with aromatic selectivity
  • Separation mechanism based on hydrophobic and π-π interactions
With the separation principle based on two retention mechanisms NUCLEODUR® Phenyl-Hexyl is an alternative to classical C18 modifications. Thus, this phase is well suited for aromatic and unsaturated compounds like aromatic acids, steroids, and flavonoids - also for LC-MS detection.
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).