Unleaded gasoline composite standards for UST monitoring
Proveedor: Restek
30081 30205 30206 30205-3 30205-5 30206-3 30206-5
REST30081EA 71.5 EUR
REST30081 REST30205 REST30206 REST30205-3 REST30205-5 REST30206-3 REST30206-5
Unleaded gasoline composite standards for UST monitoring
Normativas Estándares de la industria petrolera
Unleaded gasoline composite standards are comprised of three separate sources each of 87, 89, and 93 octanes blended in equal portions.
Gasoline standards may exhibit lot-to-lot variation.
Certificaciones: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.