Isomaltose ≥95%, crystalline solid

Proveedor: Cayman Chemical

Sinónimos: 6-O-alpha-D-Glucopyranosyl-D-glucose, (2R,3S,4R,5R)-2,3,4,5-Tetrahydroxy-6-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, alpha-1,6-Glucobiose

20797-100 20797-250 20797-50 20797-500
CAYM20797-100EA 101 EUR
CAYM20797-100 CAYM20797-250 CAYM20797-50 CAYM20797-500
Isomaltose ≥95%, crystalline solid
Isomaltose

Isomaltose is a glucose disaccharide with an α-(1 - 6) linkage, as opposed to the α-(1 - 4) linkage found in maltose. It can be liberated from dextran by dextranase and is hydrolyzed to D-glucose by isomaltase through an α-D-glucosidase-type action. Congenital sucrase-isomaltase deficiency is a rare autosomal intestinal disorder resulting from mutations affecting the gene encoding, the proprotein from which sucrase and isomaltase are produced.

Formula: C₁₂H₂₂O₁₁
Temperatura de almacenaje: Congelador
Número MDL: MFCD00065373
Núm. CAS: 499-40-1

Order Now

Resultados test especificaciones

Purity ≥95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR