Ultra-micro balances, Cubis® II series
Proveedor: Sartorius Balances
MCA2.7SF-CEU MCA2.7SF-S00 MCA2.7SM-CEU MCA2.7SM-S00
611-4693EA 44335 EUR
611-4693 611-4694 611-4695 611-4696
Ultra-micro balances, Cubis® II series
Balanzas
These ultra-micro balances from the Cubis® II range are for highly accurate weighing of microgram quantities, there are also models specifically designed for filter weighing applications.
  • Manual stainless steel draft shield for filter weighting (MCA2.75F models) or round glass motorised draft shield (MCA2.75M models)
  • Quick and easy motorised automatic levelling
  • Gesture control sensor recognises 4 gestures, it allows user to open or close the automatic draught shield, start the ioniser or send a document to a device
  • Connectivity package links to diverse laboratory information systems (Windows File Server (SMB), FTP, FTPS and STARLIMS) without the need for additional software
  • Built in ioniser removes electrostatic charges at the press of a key or automatically when draft shield closes
  • Full data integrity following ALCOA+ principles, the balances audit trail is a tamper protected, time stamped electronic logfile traceable to UTC and the balance contains an Alibi memory
Two different user interfaces are available to cover all facets of laboratory applications and enable a range of options for balance operation. The customer can then add the appropriate weighing platform to the choice of control unit. These ultra micro and micro (6 and 7 place balances) have choice of manual stainless steel draft shield for filter weighing applications or an automatic motorised round glass shield with learning capability, illumination and integrated motion sensor.
All models featue the MCA Advanced User Interface: This is a large 7” colour touch display featuring touch, scroll and swipe functionality with factory installed essential weighing applications and diverse QApp packages for optional software extension. An optional climate sensor which monitors the influencing environmental variables ensuring temperature, pressure and humidity do not negatively affect results is available. The Status Center shows all information about the balance and environmental conditions (calibration, levelling, temperature, humidity, air pressure and service) in a dashboard, giving detailed help and support in the event of warnings or errors.
Various Q-Apps, customer-specific applications, can be integrated into the balance. The applications are used to create customer-specific weighing tasks. These are reloadable application programs that guide the user step-by-step through the specific workflow, guaranteeing that the procedures described in the corresponding SOPs are observed at all times. As individual Q-Apps are performed according to their specific application, a variety of solutions for differential weighing, formula and filling quantity control applications are available as Q-App packages. Workflows can be performed independently and do not necessarily require a PC connection.
Certificaciones: Supports all requirements for FDA directive 21 CFR part II and EU Annex II compliance.
Información para pedidos: There are different optional software packages, each contains applications or functions to enhance and customise the balance for the users’ workflows. Choice of QApp Package: Pharma; Advanced Applications: Utilities; Connectivity or the all inclusive QApps package includes all of these 4 packages.
Precaución: Note: Models with suffix -CEU are verified version Europe (except FR) and -S00 suffix denotes standard version worldwide.