Ricobendazole ≥98%

Proveedor: Cayman Chemical
Atención

Sinónimos: Albendazole sulphoxide, N-[6-(propylsulphinyl)-1H-benzimidazol-2-yl]-carbamic acid, methyl ester, Albendazole Oxide, Methyl (6-(propylsulphinyl)-1H-benzo[d]imidazol-2-yl)carbamate

21880-10 21880-25 21880-5 21880-50
CAYM21880-10EA 115 EUR
CAYM21880-10 CAYM21880-25 CAYM21880-5 CAYM21880-50
Ricobendazole ≥98%
Ricobendazole

Ricobendazole, also known as albendazole sulfate, is a benzimidazole agent that inhibits tubulin polymerization and is commonly used as an antihelmintic and antiparasitic agent, with IC90 values ranging from 3.8 x 10-2 to <10-4 µg/ml against Encephalitozoon species.


  • Formulation: A crystalline solid
  • λmax: 224, 294 nm


In vitro, ricobendazole effectively kills Taenia cysts, with EC50 and EC99 values of 0.068 and 0.556 µg/ml, respectively, in 11-day cultures, and inhibits adherence in Giardia species with a 24-hour IC50 value of 3.15 µM.5,6 This agent is metabolized from albendazole via cytochrome P450 (CYP450) and may modulate CYP450 activity. In rat microsomes, ricobendazole reduces the activity of ethoxyresorufin O-deethylase (EROD) by more than 5-fold relative to albendazole, with an IC50 value of 5 µM.

Formula: C₁₂H₁₅N₃O₃S
MW: 281,3 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00797922
Núm. CAS: 54029-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR