Taurodeoxycholic acid sodium salt ≥95%
Proveedor: Cayman Chemical
|
Atención
|
15935-25
15935-5
15935-1
CAYM15935-25EA
622
EUR
CAYM15935-25
CAYM15935-5
CAYM15935-1
Taurodeoxycholic acid sodium salt ≥95%
Taurodeoxycholic acid sodium salt
Taurodeoxycholic acid is a taurine-conjugated form of the secondary bile acid deoxycholic acid. Taurodeoxycholic acid stimulates chloride ion secretion through calcium-activated chloride ion channels and cystic fibrosis transmembrane conductance regulator (CFTR) in Calu-3 airway epithelial cell monolayers when applied basolaterally. Serum levels of taurodeoxycholic acid increase approximately 5-fold in within two hours during an oral lipid tolerance test in humans.
Formula:
C₂₆H₄₄NO₆S·Na |
Número MDL:
MFCD00003671 Núm. CAS: 1180-95-6 |
Resultados test especificaciones
Purity | ≥95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...