Organic reference standard, Ammelide, analytical standard, Supelco®
Proveedor: Merck
49866-10MGEA 83.1 EUR
49866-10MG
Organic reference standard, Ammelide, analytical standard, Supelco®
Normativas Estándares orgánicos
Ammelide can be synthesised via hydrolysis of 2-amino-4,6-dichloro-s-triazine in the presence of acid or base or by the reaction between urea and cyanogen iodide at 130 to 140 degrees.
Envase: Bottomless glass bottle contents are inside inserted with fused cone.