Organic reference standard, gamma-Butyrolactone, analytical standard, Supelco®
Proveedor: Merck
90970-1ML
90970-5ML
90970-1MLEA
57.1
EUR
90970-1ML
90970-5ML
Organic reference standard, gamma-Butyrolactone, analytical standard, Supelco®
Normativas
Estándares orgánicos
Precursor of γ-hydroxybutyric acid (GHB). It blocks dopamine release by blocking impulse flow in dopaminergic neurons. Pretreatment with γ-butyrolactone allows detection of autoreceptor-induced dopamine release.