Organic reference standard, gamma-Butyrolactone, analytical standard, Supelco®
Proveedor: Merck
90970-1ML 90970-5ML
90970-1MLEA 57.1 EUR
90970-1ML 90970-5ML
Organic reference standard, gamma-Butyrolactone, analytical standard, Supelco®
Normativas Estándares orgánicos
Precursor of γ-hydroxybutyric acid (GHB). It blocks dopamine release by blocking impulse flow in dopaminergic neurons. Pretreatment with γ-butyrolactone allows detection of autoreceptor-induced dopamine release.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba81a586bca1b2330a5e59af649497809a16b3a&vid=4ba81a586bca1b2330a5e59af649497809a16b3a&prid=web&clid=SAPDAM
Order Now
ESPECIFICACIONES
Descripción gamma-Butyrolactone
Environmentally Preferable
CAS No. 96-48-0
Formula C₄H₆O₂
Melting point −45 °C
Punto de ebullición 204...205 °C
Temperatura de almacenamiento 2..8 °C