Colour reference solutions acc. to Ph Eur, analytical standard, Set of colours B, BY, Y, GY, R
Proveedor: Merck
90232-1SET-FEA 1140 EUR
90232-1SET-F 83952-1SET-F
Colour reference solutions acc. to Ph Eur, analytical standard, Set of colours B, BY, Y, GY, R
Normativas Patrones de espectroscopia
This is a set of the following color reference solutions sets according to Ph. Eur.:
Colour Reference Solutions B acc. to PhEur - 10×2 ml ampoules or 10×10 ml ampoules
Colour Reference Solutions BY acc. to PhEur - 8×2 ml ampoules or 8×10 ml ampoles
Colour Reference Solutions Y acc. to PhEur - 8×2 ml ampoules or 8×10 ml ampoles
Colour Reference Solutions GY acc. to PhEur - 8×2 ml ampoules or 8×10 ml ampoles
Colour Reference Solutions R acc. to PhEur - 8×2 ml ampoules or 8×10 ml ampoles