Tetrabutylammonium benzoate ≥99.0% (by titrimetric analysis) for electrochemical analysis, Supelco®

Proveedor: Merck
Atención


M-Clarity™ quality level = MQ100

86837-25G 86837-5G
86837-25GEA 478 EUR
86837-25G 86837-5G
Tetrabutylammonium benzoate ≥99.0% (by titrimetric analysis) for electrochemical analysis, Supelco®
Tetrabutylammonium benzoate
Formula: C₂₃H₄₂NO₂
Número MDL: MFCD00171701
Núm. CAS: 18819-89-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70a38059a0da08fdefdef16aa3d58a41dfbf3b72&vid=778be9ba498b49108c9d7bb8e8bc7f8eac483297&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR