1,2-Epoxyheptane

Proveedor: TCI
TCIAE0312-5MLEA 238 EUR
TCIAE0312-5ML
1,2-Epoxyheptane
Normativas Estándares orgánicos
Specific gravity (20/20): 0,8350 to 0,8400
Refractive index n20/D: 1,4130 to 1,4150
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR