Guard columns, TCI OPTI-GUARD™ Fit
Proveedor: TCI
S1440-3CLM S1441-3CLM S1442-3CLM S1443-3CLM S1444-3CLM S1445-3CLM S1446-3CLM S1447-3CLM S1453-3CLM
TCIAS1440-3CLMEA 144 EUR
TCIAS1440-3CLM TCIAS1441-3CLM TCIAS1442-3CLM TCIAS1443-3CLM TCIAS1444-3CLM TCIAS1445-3CLM TCIAS1446-3CLM TCIAS1447-3CLM TCIAS1453-3CLM
Guard columns, TCI OPTI-GUARD™ Fit
Columnas de cromatografía
The TCI OPTI-GUARD™ series are compact, high-performance, and easy-to-use HPLC guard columns.
  • Special fitting style
  • Easy hand tight connection
  • For LC-MS analysis
This series is recommended for the further purification of relatively clean samples using either 4,6 or 2,0 mm I.Ø HPLC columns.