Ioduro de calcio hidrato ≥99.9965% (metals basis), Puratronic®
Proveedor: Thermo Scientific
|
Atención
|
11117.06
11117.14
11117.06EA
71.3
EUR
11117.06
11117.14
Ioduro de calcio hidrato ≥99.9965% (metals basis), Puratronic®
Ioduro de calcio hidrato
Purity: 99.9965%
CAS No.: 71626-98-7
Molecular Formula: CaI2.xH2O (x≈6)
Formula Weight: 293.89(anhy)
Physical Form: Crystalline
Density: 2.55
Appearance: Yellow
Melting Point: 42° dec.
Solubility: Freely soluble in water and alcohol
Merck Reference: 14,1677
MDL No.: MFCD00149622
Formula:
CaI₂·H₂O MW: 311,9 g/mol Pto. de fusión: 42 °C (desc.) Densidad: 2,55 Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00149622 Núm. CAS: 71626-98-7 EINECS: 233-276-8 Índice Merck: 14,1677 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...