Alloxan monohydrate ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Pyrimidine-2,4,5,6(1H,3H)-tetraone hydrate
A0216-25G
A0216-5G
TCIAA0216-25GEA
85
EUR
TCIAA0216-25G
TCIAA0216-5G
Alloxan monohydrate ≥98.0% (by titrimetric analysis)
Alloxan monohydrate
Formula:
C₄H₄N₂O₅ MW: 160,09 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00006031 Núm. CAS: 2244-11-3 Índice Merck: 14,00282 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Solubility in Water | almost transparency |
Water | 10.5 to 12.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...