5-Aminolevulinic acid hydrochloride ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: 5-Amino-4-ketovaleric acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, δ-Aminolevulinic Acid Hydrochloride

A0325-1G A0325-100MG
TCIAA0325-1GEA 53 EUR
TCIAA0325-1G TCIAA0325-100MG
5-Aminolevulinic acid hydrochloride ≥98.0% (by titrimetric analysis)
5-Aminolevulinic acid hydrochloride
Formula: C₅H₁₀ClNO₃
MW: 167.59 g/mol
Número MDL: MFCD00012869
Núm. CAS: 5451-09-2
EINECS: 226-679-5
Índice Merck: 14,00446

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR