6-Amino-2-naphthalenesulphonic acid monohydrate ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
Bronner's acid, 2-Naphthylamine-6-sulphonic acid
A0348-25G
A0348-500G
TCIAA0348-25GEA
40
EUR
TCIAA0348-25G
TCIAA0348-500G
6-Amino-2-naphthalenesulphonic acid monohydrate ≥98.0% (by HPLC)
6-Amino-2-naphthalenesulphonic acid monohydrate
Formula:
C₁₀H₉NO₃S MW: 223,25 g/mol |
Número MDL:
MFCD04038015 Núm. CAS: 93-00-5 EINECS: 202-208-9 |
Resultados test especificaciones
Appearance | White to Light red to Green powder to crystalline |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 95.0 % (calcd.on anh.substance) |
Water | 7.1 to 8.1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...