1-Amino-8-naphthol-2,4-disulphonic acid monosodium salt hydrate ≥85.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
SS acid, Monosodium 1-Amino-8-naphthol-2,4-disulphonate, 2S acid, Chicago acid, 4-Amino-5-hydroxy-1,3-naphthalenedisulphonic acid Monosodium salt, Sodium Hydrogen 4-Amino-5-hydroxy-1,3-naphthalenedisulphonate
A0363-25G
A0363-500G
TCIAA0363-25GEA
96
EUR
TCIAA0363-25G
TCIAA0363-500G
1-Amino-8-naphthol-2,4-disulphonic acid monosodium salt hydrate ≥85.0% (by HPLC, titration analysis)
1-Amino-8-naphthol-2,4-disulphonic acid monosodium salt hydrate
Formula:
C₁₀H₉NO₇S₂ MW: 341.28 g/mol |
Número MDL:
MFCD00068275 Núm. CAS: 52789-62-5 EINECS: 258-185-0 |
Resultados test especificaciones
Appearance | Light gray to Brown to Black powder to crystal |
Purity (HPLC) | min. 85.0 area% |
Purity (Neutralization titration) | min. 85.0 % (calcd.on anh.substance) |
Water | max. 10.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...