6-Amino-1-naphthol-3-sulphonic acid hydrate ≥95.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
7-Amino-4-hydroxy-2-naphthalenesulfonic acid, 7-Amino-4-hydroxy-2-naphthalenesulphonic acid, I-acid, J acid, 7-Amino-4-hydroxynaphthalene-2-sulphonic acid, 6-Amino-1-naphthol-3-sulphonic Acid
A0370-25G
A0370-500G
TCIAA0370-25GEA
16
EUR
TCIAA0370-25G
TCIAA0370-500G
6-Amino-1-naphthol-3-sulphonic acid hydrate ≥95.0% (by HPLC, titration analysis)
6-Amino-1-naphthol-3-sulphonic acid hydrate
Formula:
C₁₀H₉NO₄S MW: 239,25 g/mol |
Número MDL:
MFCD00003970 Núm. CAS: 87-02-5 EINECS: 201-718-9 |
Resultados test especificaciones
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 95.0 % (calcd.on anh.substance) |
Water | max. 8.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...