D-Amygdalin ≥97.0% (by HPLC)

Proveedor: TCI
Atención

Sinónimos: Laetrile, Amygdalin, Amygdaloside, D-Mandelonitrile 6-O-β-D-glucosido-β-D-glucoside, (R)-2-Phenyl-2-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)acetonitrile, D-Mandelonitrile-β-gentiobioside

A0443-1G A0443-10G A0443-5G
TCIAA0443-1GEA 16 EUR
TCIAA0443-1G TCIAA0443-10G TCIAA0443-5G
D-Amygdalin ≥97.0% (by HPLC)
D-Amygdalin
Formula: C₂₀H₂₇NO₁₁
MW: 457,43 g/mol
Número MDL: MFCD00006598
Núm. CAS: 29883-15-6
EINECS: 249-925-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00aa56199985fa0812da7658ea8f8fc3117920d9&vid=06b6db2651dfb964ea8903922e3c43ee0283d755&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 93.0 %
Specific rotation [a]20/D -35.0 to -45.0 deg (C=2, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR