DL-Asparagine monohydrate ≥99.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
DL-Aspartic acid 4-amide monohydrate, H-DL-Asn-OH monohydrate, (±)-2-Aminosuccinic acid 4-amide monohydrate, DL-Asparagine hydrate, H-DL-Asn-OH·H₂O, DL-2-Aminosuccinamic acid monohydrate, 2,4-Diamino-4-oxobutanoic acid monohydrate
A0541-25G
A0541-500G
A0541-100G
TCIAA0541-25GEA
23
EUR
TCIAA0541-25G
TCIAA0541-500G
TCIAA0541-100G
DL-Asparagine monohydrate ≥99.0% (by titrimetric analysis)
DL-Asparagine monohydrate
Formula:
C₄H₈N₂O₃·H₂O MW: 150,13 g/mol |
Número MDL:
MFCD00151039 Núm. CAS: 3130-87-8 EINECS: 221-521-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 99.0% (calcd.on dried substance) |
Drying loss | 11.5 to 12.7 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...