Azo Blue
Proveedor: TCI
A0587-25G
A0587-500G
TCIAA0587-25GEA
38
EUR
TCIAA0587-25G
TCIAA0587-500G
Azo Blue
Azo Blue
Formula:
C₃₄H₂₆N₄O₈S₂ |
Número MDL:
MFCD00059133 Núm. CAS: 6059-34-3 |
|
Resultados test especificaciones
Appearance | Black powder to crystal |
Lambda max. | 555.0 to 560.0 nm (methanol) |
Absorbance (E1%1cm) | min. 600 (methanol, 555.0 to 560.0 nm) |
Drying loss | max. 12.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...