1-Adamantaneacetic acid ≥99.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-(Adamantan-1-yl)acetic acid, 1-Carboxymethyladamantane
A0736-25G
A0736-5G
TCIAA0736-25GEA
202
EUR
TCIAA0736-25G
TCIAA0736-5G
1-Adamantaneacetic acid ≥99.0% (by titrimetric analysis)
1-Adamantaneacetic acid
Formula:
C₁₂H₁₈O₂ MW: 194.27 g/mol |
Número MDL:
MFCD00074728 Núm. CAS: 4942-47-6 EINECS: 225-585-1 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 135.0 to 139.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...